siRNA Details
| Gene Symbol: Usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015015.1 | NP_001015015.1 | 9 | 2378 | ||
| GenBank mRNA | BC092574.1 | - | 9 | 2378 | 7113947 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp1| Gene Symbol: Usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015015.1 | NP_001015015.1 | 9 | 2378 | ||
| GenBank mRNA | BC092574.1 | - | 9 | 2378 | 7113947 | |