siRNA Details
| Gene Symbol: Dnajb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015021.1 | NP_001015021.1 | 10 | 1806 | ||
| GenBank mRNA | BC093384.1 | - | 10 | 1806 | 7389370 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajb11| Gene Symbol: Dnajb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015021.1 | NP_001015021.1 | 10 | 1806 | ||
| GenBank mRNA | BC093384.1 | - | 10 | 1806 | 7389370 | |