siRNA Details
| Gene Symbol: Tbc1d10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015022.3 | NP_001015022.1 | 7 | 785 | ||
| GenBank mRNA | AB210856.1 | - | 7 | 785 | ||
| BC093603.1 | - | 7 | 848 | 7316341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d10a| Gene Symbol: Tbc1d10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015022.3 | NP_001015022.1 | 7 | 785 | ||
| GenBank mRNA | AB210856.1 | - | 7 | 785 | ||
| BC093603.1 | - | 7 | 848 | 7316341 | ||