siRNA Details
| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017378.1 | NP_001017378.1 | 3 | 334 | ||
| GenBank mRNA | BC083891.1 | - | 3 | 334 | 7132551 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kat8| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017378.1 | NP_001017378.1 | 3 | 334 | ||
| GenBank mRNA | BC083891.1 | - | 3 | 334 | 7132551 | |