siRNA Details
| Gene Symbol: Kdm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017426.1 | NP_001017426.1 | 5 | 628 | ||
| GenBank mRNA | BC075632.1 | - | 6 | 804 | 30362899 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdm6b| Gene Symbol: Kdm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017426.1 | NP_001017426.1 | 5 | 628 | ||
| GenBank mRNA | BC075632.1 | - | 6 | 804 | 30362899 | |