siRNA Details
Gene Symbol: Rasef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017427.1 | NP_001017427.1 | 3 | 378 | ||
GenBank mRNA | AK132691.1 | - | 3 | 358 | ||
BB616033.1 | - | 3 | 380 | |||
BC125539.1 | - | 2 | 192 | 40130054 | ||
BC137774.1 | - | 2 | 192 | 8860793 | ||
BC144901.1 | - | 2 | 192 |
