siRNA Details
| Gene Symbol: Rasef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017427.1 | NP_001017427.1 | 9 | 955 | ||
| GenBank mRNA | AK132691.1 | - | 9 | 935 | ||
| BC030882.1 | - | 5 | 352 | |||
| BC040766.1 | - | 5 | 341 | |||
| BC125539.1 | - | 8 | 769 | 40130054 | ||
| BC137774.1 | - | 8 | 769 | 8860793 | ||
| BC144901.1 | - | 8 | 769 | |||

Rasef
Genomic Map