siRNA Details
| Gene Symbol: Nsun7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017452.1 | NP_001017452.1 | 4, 5 | 789 | ||
| XM_006250946.2 | XP_006251008.1 | 4, 5 | 1080 | |||
| XM_006250947.2 | XP_006251009.1 | 4, 5 | 1083 | |||
| XM_006250948.2 | XP_006251010.1 | 4, 5 | 1080 | |||
| GenBank mRNA | BC083653.1 | - | 4, 5 | 789 | 7135159 | |

Nsun7
Genomic Map