siRNA Details
| Gene Symbol: Ist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017454.2 | NP_001017454.1 | 10 | 1536 | ||
| XM_008772506.1 | XP_008770728.1 | 10 | 1503 | |||
| XM_008772507.1 | XP_008770729.1 | 9 | 1452 | |||
| XM_008772508.1 | XP_008770730.1 | 8 | 1242 | |||
| GenBank mRNA | BC092631.1 | - | 10 | 1493 | 7314864 | |

Ist1
Genomic Map