siRNA Details
| Gene Symbol: VMA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017980.3 | NP_001017980.1 | 3 | 4565 | ||
| XM_011531125.2 | XP_011529427.1 | 3 | 4891 | |||
| GenBank mRNA | AF086459.1 | - | 1 | 408 | ||
| AK025798.1 | - | 1 | 1913 | |||
| AK091301.1 | - | 1 | 3273 | |||
| AL833596.1 | - | 3 | 4518 | |||
| BC110800.1 | - | 3 | 4479 | 6044374 | ||

VMA21
Genomic Map