siRNA Details
| Gene Symbol: Gm4983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC030401.1 | - | 1400 | 5362444 | ||
| BC083083.1 | - | 1258 | 6514154 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4983| Gene Symbol: Gm4983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC030401.1 | - | 1400 | 5362444 | ||
| BC083083.1 | - | 1258 | 6514154 | |||