siRNA Details
| Gene Symbol: Taok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024254.1 | NP_001019425.1 | 1785 | |||
| GenBank mRNA | AB195232.1 | - | 1785 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taok3| Gene Symbol: Taok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024254.1 | NP_001019425.1 | 1785 | |||
| GenBank mRNA | AB195232.1 | - | 1785 | |||