siRNA Details
| Gene Symbol: Tspan11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024262.1 | NP_001019433.1 | 8 | 2363 | ||
| GenBank mRNA | BC092652.1 | - | 8 | 2363 | 7320221 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan11| Gene Symbol: Tspan11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024262.1 | NP_001019433.1 | 8 | 2363 | ||
| GenBank mRNA | BC092652.1 | - | 8 | 2363 | 7320221 | |