siRNA Details
| Gene Symbol: Rnf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024271.1 | NP_001019442.1 | 7 | 1246 | ||
| XM_006251333.1 | XP_006251395.1 | 7 | 1214 | |||
| XM_008770331.1 | XP_008768553.1 | 5 | 1058 | |||
| GenBank mRNA | BC092655.1 | - | 7 | 1246 | 7320514 | |

Rnf185
Genomic Map