siRNA Details
| Gene Symbol: Snx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024560.2 | NP_001019731.1 | 13 | 1645 | ||
| GenBank mRNA | BC048922.1 | - | 13 | 1645 | 6311864 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx32| Gene Symbol: Snx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024560.2 | NP_001019731.1 | 13 | 1645 | ||
| GenBank mRNA | BC048922.1 | - | 13 | 1645 | 6311864 | |