siRNA Details
| Gene Symbol: Ttll10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024758.2 | NP_001019929.1 | 9 | 1037 | ||
| NM_001170533.1 | NP_001164004.1 | 10 | 1459 | |||
| XM_006239559.2 | XP_006239621.1 | 10 | 1619 | |||
| XM_006239562.2 | XP_006239624.1 | 8 | 1267 | |||
| XM_008764356.1 | XP_008762578.1 | 11 | 2906 | |||
| XM_008764357.1 | XP_008762579.1 | 10 | 1415 | |||
| GenBank mRNA | BC083833.1 | - | 7 | 876 | ||
| BC097417.1 | - | 9 | 1037 | 7454407 | ||

Ttll10
Genomic Map