siRNA Details
| Gene Symbol: Wdr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024766.1 | NP_001019937.1 | 1 | 1225 | ||
| GenBank mRNA | BC097449.1 | - | 1 | 1225 | 7456711 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr5b| Gene Symbol: Wdr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024766.1 | NP_001019937.1 | 1 | 1225 | ||
| GenBank mRNA | BC097449.1 | - | 1 | 1225 | 7456711 | |