siRNA Details
Gene Symbol: Tnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024771.1 | NP_001019942.1 | 5 | 1839 | ||
XM_006251255.2 | XP_006251317.1 | 6 | 1993 | |||
GenBank mRNA | BC085912.1 | - | 5 | 1770 | ||
BC097956.1 | - | 5 | 1839 | 7461734 |
