siRNA Details
| Gene Symbol: Zfp110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024775.1 | NP_001019946.1 | 8 | 1253 | ||
| XM_006228111.2 | XP_006228173.1 | 7 | 1984 | |||
| XM_006228112.2 | XP_006228174.1 | 8 | 1817 | |||
| GenBank mRNA | BC097420.1 | - | 8 | 1253 | 7454819 | |

Zfp110
Genomic Map