siRNA Details
| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024776.1 | NP_001019947.1 | 4 | 2274 | ||
| GenBank mRNA | BC098070.1 | - | 4 | 2274 | 7455473 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hipk4| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024776.1 | NP_001019947.1 | 4 | 2274 | ||
| GenBank mRNA | BC098070.1 | - | 4 | 2274 | 7455473 | |