siRNA Details
| Gene Symbol: Pdzd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759743.1 | XP_008757965.1 | 4 | 1893 | ||
| GenBank mRNA | BC097426.1 | - | 4 | 1879 | 7455141 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdzd9| Gene Symbol: Pdzd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759743.1 | XP_008757965.1 | 4 | 1893 | ||
| GenBank mRNA | BC097426.1 | - | 4 | 1879 | 7455141 | |