siRNA Details
| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024778.1 | NP_001019949.1 | 7, 8 | 936 | ||
| XM_006231848.2 | XP_006231910.1 | 7, 8 | 932 | |||
| GenBank mRNA | BC098057.1 | - | 7, 8 | 936 | 7457767 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad17| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024778.1 | NP_001019949.1 | 7, 8 | 936 | ||
| XM_006231848.2 | XP_006231910.1 | 7, 8 | 932 | |||
| GenBank mRNA | BC098057.1 | - | 7, 8 | 936 | 7457767 | |