siRNA Details
| Gene Symbol: Actl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024788.1 | NP_001019959.1 | 1 | 3241 | ||
| GenBank mRNA | AY635905.1 | - | 1 | 3241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actl11| Gene Symbol: Actl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024788.1 | NP_001019959.1 | 1 | 3241 | ||
| GenBank mRNA | AY635905.1 | - | 1 | 3241 | ||