siRNA Details
Gene Symbol: Actl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024788.1 | NP_001019959.1 | 1 | 3517 | ||
GenBank mRNA | AY635905.1 | - | 1 | 3517 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
