siRNA Details
| Gene Symbol: Usp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024790.1 | NP_001019961.1 | 30 | 3194 | ||
| XM_006245756.2 | XP_006245818.1 | 31 | 3406 | |||
| XM_006245757.2 | XP_006245819.1 | 30 | 4243 | |||
| XM_006245758.2 | XP_006245820.1 | 30 | 3336 | |||
| XM_006245759.1 | XP_006245821.1 | 30 | 3287 | |||
| GenBank mRNA | AY641530.1 | - | 30 | 3194 | ||

Usp7
Genomic Map