siRNA Details
| Gene Symbol: Epn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024791.1 | NP_001019962.1 | 9 | 3430 | ||
| XM_006247201.2 | XP_006247263.1 | 10 | 4614 | |||
| XM_006247202.2 | XP_006247264.1 | 10 | 4579 | |||
| XM_006247203.2 | XP_006247265.1 | 9 | 4533 | |||
| GenBank mRNA | BC097500.1 | - | 9 | 3430 | 7462750 | |

Epn3
Genomic Map