siRNA Details
| Gene Symbol: LMBR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300750.1 | NP_001287679.1 | 7 | 1095 | ||
| NM_001300751.1 | NP_001287680.1 | 6 | 893 | |||
| NM_018113.3 | NP_060583.2 | 6 | 893 | |||
| XM_011538563.2 | XP_011536865.1 | 6 | 1047 | |||
| XM_011538564.2 | XP_011536866.1 | 6 | 1420 | |||
| XM_011538565.2 | XP_011536867.1 | 5 | 1669 | |||
| XM_011538566.2 | XP_011536868.1 | 6 | 708 | |||
| XM_017019626.1 | XP_016875115.1 | 6 | 1295 | |||
| XM_017019627.1 | XP_016875116.1 | 7 | 1084 | |||
| XM_017019628.1 | XP_016875117.1 | 6 | 882 | |||
| XM_017019629.1 | XP_016875118.1 | 7 | 1939 | |||
| XM_017019630.1 | XP_016875119.1 | 4 | 2920 | |||
| XM_017019631.1 | XP_016875120.1 | 6 | 708 | |||
| GenBank mRNA | AB033000.1 | - | 6 | 660 | ||
| AF260728.1 | - | 6 | 852 | |||
| AK001356.1 | - | 7 | 876 | |||
| AK001651.1 | - | 6 | 843 | |||
| AK093570.1 | - | 1 | 679 | |||
| AK298026.1 | - | 5 | 836 | |||
| AK310064.1 | - | 7 | 1029 | |||
| AL137599.1 | - | 3 | 286 | |||
| AL556633.3 | - | 6 | 857 | |||
| AY358572.1 | - | 6 | 857 | |||
| BC008337.1 | - | 6 | 823 | 3506347 | ||
| BC015015.1 | - | 6 | 622 | 3910520 | ||
| BC031550.1 | - | 6 | 654 | 5194023 | ||
| KU178711.1 | - | 6 | 548 | |||
| KU178712.1 | - | 5 | 531 | |||
| KU178713.1 | - | 5 | 514 | |||
| KU178714.1 | - | 6 | 548 | |||
| KU178715.1 | - | 6 | 548 | |||

LMBR1L
Genomic Map