siRNA Details
| Gene Symbol: Cdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024793.1 | NP_001019964.1 | 12 | 1601 | ||
| XM_006247533.2 | XP_006247595.1 | 11 | 3322 | |||
| XM_006247534.2 | XP_006247596.1 | 11 | 3319 | |||
| XM_006247535.2 | XP_006247597.1 | 12 | 1738 | |||
| XM_006247536.2 | XP_006247598.1 | 11 | 3323 | |||
| XM_006247537.1 | XP_006247599.1 | 9 | 1270 | |||
| XM_008768295.1 | XP_008766517.1 | 11 | 1591 | |||
| GenBank mRNA | BC097475.1 | - | 12 | 1601 | 7457819 | |

Cdc27
Genomic Map