siRNA Details
| Gene Symbol: Cdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024793.1 | NP_001019964.1 | 12, 13 | 1633 | ||
| XM_006247533.2 | XP_006247595.1 | 11, 12 | 3354 | |||
| XM_006247534.2 | XP_006247596.1 | 11, 12 | 3351 | |||
| XM_006247535.2 | XP_006247597.1 | 12, 13 | 1770 | |||
| XM_006247537.1 | XP_006247599.1 | 9, 10 | 1302 | |||
| XM_008768295.1 | XP_008766517.1 | 11, 12 | 1623 | |||
| GenBank mRNA | BC097475.1 | - | 12, 13 | 1633 | 7457819 | |

Cdc27
Genomic Map