siRNA Details
| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024794.1 | NP_001019965.1 | 4 | 818 | ||
| XM_008770555.1 | XP_008768777.1 | 4 | 1191 | |||
| GenBank mRNA | BC097400.1 | - | 4 | 818 | 7453630 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl3| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024794.1 | NP_001019965.1 | 4 | 818 | ||
| XM_008770555.1 | XP_008768777.1 | 4 | 1191 | |||
| GenBank mRNA | BC097400.1 | - | 4 | 818 | 7453630 | |