siRNA Details
| Gene Symbol: Gpalpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024875.1 | NP_001020046.1 | 5 | 599 | ||
| GenBank mRNA | BC097485.1 | - | 5 | 599 | 7459542 | |
| FQ214079.1 | - | 287 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpalpp1| Gene Symbol: Gpalpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024875.1 | NP_001020046.1 | 5 | 599 | ||
| GenBank mRNA | BC097485.1 | - | 5 | 599 | 7459542 | |
| FQ214079.1 | - | 287 | ||||