siRNA Details
| Gene Symbol: Ccdc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024882.1 | NP_001020053.1 | 7 | 814 | ||
| XM_006250296.2 | XP_006250358.1 | 7 | 909 | |||
| GenBank mRNA | BC098058.1 | - | 7 | 814 | 7457890 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc19| Gene Symbol: Ccdc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024882.1 | NP_001020053.1 | 7 | 814 | ||
| XM_006250296.2 | XP_006250358.1 | 7 | 909 | |||
| GenBank mRNA | BC098058.1 | - | 7 | 814 | 7457890 | |