siRNA Details
| Gene Symbol: Pomk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024883.1 | NP_001020054.1 | 5 | 1330 | ||
| XM_008771353.1 | XP_008769575.1 | 3 | 1104 | |||
| GenBank mRNA | BC097466.1 | - | 5 | 1330 | 7457667 | |
| FQ211927.1 | - | 5 | 1319 | |||

Pomk
Genomic Map