siRNA Details
| Gene Symbol: Ccdc90b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024885.1 | NP_001020056.1 | 1, 2 | 410 | ||
| GenBank mRNA | BC097480.1 | - | 1, 2 | 410 | 7458611 | |
| FQ215669.1 | - | 1, 2 | 112 | |||
| FQ216256.1 | - | 1, 2 | 114 | |||
| FQ218235.1 | - | 1, 2 | 125 | |||

Ccdc90b
Genomic Map