siRNA Details
| Gene Symbol: Sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024887.1 | NP_001020058.1 | 10 | 1453 | ||
| XM_006230552.2 | XP_006230614.1 | 10 | 2287 | |||
| XM_006230553.2 | XP_006230615.1 | 10 | 1919 | |||
| XM_008759988.1 | XP_008758210.1 | 9 | 3485 | |||
| XM_008759989.1 | XP_008758211.1 | 10 | 1978 | |||
| XM_008759990.1 | XP_008758212.1 | 10 | 1481 | |||
| GenBank mRNA | BC097488.1 | - | 10 | 1453 | 7460323 | |

Sigirr
Genomic Map