siRNA Details
| Gene Symbol: Sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024887.1 | NP_001020058.1 | 5 | 640 | ||
| XM_006230552.2 | XP_006230614.1 | 5 | 1474 | |||
| XM_006230553.2 | XP_006230615.1 | 5 | 1106 | |||
| XM_008759988.1 | XP_008758210.1 | 4 | 2672 | |||
| XM_008759989.1 | XP_008758211.1 | 5 | 1165 | |||
| XM_008759990.1 | XP_008758212.1 | 5 | 668 | |||
| GenBank mRNA | BC097488.1 | - | 5 | 640 | 7460323 | |

Sigirr
Genomic Map