siRNA Details
| Gene Symbol: Gatad2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024888.1 | NP_001020059.1 | 7 | 1357 | ||
| XM_008761144.1 | XP_008759366.1 | 7 | 1288 | |||
| XM_008761145.1 | XP_008759367.1 | 7 | 1285 | |||
| GenBank mRNA | BC097428.1 | - | 7 | 1357 | 7455294 | |

Gatad2b
Genomic Map