siRNA Details
| Gene Symbol: Pih1d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024890.1 | NP_001020061.1 | 3 | 179 | ||
| GenBank mRNA | BC097458.1 | - | 3 | 179 | 7457221 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pih1d3| Gene Symbol: Pih1d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024890.1 | NP_001020061.1 | 3 | 179 | ||
| GenBank mRNA | BC097458.1 | - | 3 | 179 | 7457221 | |