siRNA Details
| Gene Symbol: Trp53i13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024920.1 | NP_001020091.1 | 6 | 1042 | ||
| GenBank mRNA | AK003215.1 | - | 1 | 312 | ||
| BC062271.1 | - | 1 | 271 | |||
| BC147324.1 | - | 6 | 1040 | 9056369 | ||
| BC147325.1 | - | 6 | 1040 | 9056377 | ||

Trp53i13
Genomic Map