siRNA Details
| Gene Symbol: Ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024922.2 | NP_001020093.2 | 8 | 913 | ||
| GenBank mRNA | BC085301.1 | - | 8 | 913 | ||
| BC099554.1 | - | 8 | 920 | 30308124 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx49| Gene Symbol: Ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024922.2 | NP_001020093.2 | 8 | 913 | ||
| GenBank mRNA | BC085301.1 | - | 8 | 913 | ||
| BC099554.1 | - | 8 | 920 | 30308124 | ||