siRNA Details
| Gene Symbol: Exoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024964.1 | NP_001020135.1 | 5, 6 | 1246 | ||
| XM_006227767.2 | XP_006227829.1 | 5, 6 | 1364 | |||
| XM_006227768.2 | XP_006227830.1 | 5, 6 | 1660 | |||
| GenBank mRNA | BC097993.1 | - | 5, 6 | 1246 | 7446619 | |
| U32575.1 | - | 4, 5 | 1201 | |||

Exoc3
Genomic Map