siRNA Details
| Gene Symbol: Nrros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024995.1 | NP_001020166.1 | 3 | 1592 | ||
| XM_006248448.2 | XP_006248510.1 | 3 | 1633 | |||
| GenBank mRNA | BC091190.1 | - | 3 | 1592 | 7367889 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrros| Gene Symbol: Nrros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024995.1 | NP_001020166.1 | 3 | 1592 | ||
| XM_006248448.2 | XP_006248510.1 | 3 | 1633 | |||
| GenBank mRNA | BC091190.1 | - | 3 | 1592 | 7367889 | |