siRNA Details
| Gene Symbol: Txndc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024998.1 | NP_001020169.1 | 3 | 700 | ||
| XM_008771497.1 | XP_008769719.1 | 3 | 808 | |||
| GenBank mRNA | BC091340.1 | - | 3 | 700 | 7315216 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txndc15| Gene Symbol: Txndc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024998.1 | NP_001020169.1 | 3 | 700 | ||
| XM_008771497.1 | XP_008769719.1 | 3 | 808 | |||
| GenBank mRNA | BC091340.1 | - | 3 | 700 | 7315216 | |