siRNA Details
| Gene Symbol: Klhl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025008.1 | NP_001020179.1 | 4 | 2252 | ||
| XM_008765509.1 | XP_008763731.1 | 4 | 2433 | |||
| GenBank mRNA | BC091195.1 | - | 4 | 2252 | 7374210 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl38| Gene Symbol: Klhl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025008.1 | NP_001020179.1 | 4 | 2252 | ||
| XM_008765509.1 | XP_008763731.1 | 4 | 2433 | |||
| GenBank mRNA | BC091195.1 | - | 4 | 2252 | 7374210 | |