siRNA Details
| Gene Symbol: Josd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025009.1 | NP_001020180.1 | 4 | 1058 | ||
| GenBank mRNA | BC091280.1 | - | 4 | 1058 | 7384518 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Josd1| Gene Symbol: Josd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025009.1 | NP_001020180.1 | 4 | 1058 | ||
| GenBank mRNA | BC091280.1 | - | 4 | 1058 | 7384518 | |