siRNA Details
Gene Symbol: Josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025009.1 | NP_001020180.1 | 4 | 1730 | ||
GenBank mRNA | BC091280.1 | - | 4 | 1730 | 7384518 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025009.1 | NP_001020180.1 | 4 | 1730 | ||
GenBank mRNA | BC091280.1 | - | 4 | 1730 | 7384518 |