siRNA Details
| Gene Symbol: RGD1305464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025011.2 | NP_001020182.2 | 3 | 3480 | ||
| GenBank mRNA | BC089935.1 | - | 2 | 554 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305464| Gene Symbol: RGD1305464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025011.2 | NP_001020182.2 | 3 | 3480 | ||
| GenBank mRNA | BC089935.1 | - | 2 | 554 | ||