siRNA Details
| Gene Symbol: KIF26B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018012.3 | NP_060482.2 | 13, 14 | 6475 | ||
| GenBank mRNA | AK001019.1 | - | 2, 3 | 1414 | ||
| AK125187.1 | - | 6, 7 | 1930 | |||
| AY923834.1 | - | 13, 14 | 6476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF26B| Gene Symbol: KIF26B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018012.3 | NP_060482.2 | 13, 14 | 6475 | ||
| GenBank mRNA | AK001019.1 | - | 2, 3 | 1414 | ||
| AK125187.1 | - | 6, 7 | 1930 | |||
| AY923834.1 | - | 13, 14 | 6476 | |||