siRNA Details
Gene Symbol: Tmem50b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025014.1 | NP_001020185.1 | 7 | 1626 | ||
XM_006248066.2 | XP_006248128.1 | 8 | 1794 | |||
XM_006248067.2 | XP_006248129.1 | 8 | 1880 | |||
XM_006248068.2 | XP_006248130.1 | 7 | 1746 | |||
XM_006248069.2 | XP_006248131.1 | 7 | 1632 | |||
GenBank mRNA | BC091349.1 | - | 7 | 1626 | 7316764 |
