siRNA Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025015.1 | NP_001020186.1 | 2 | 379 | ||
| GenBank mRNA | BC091236.1 | - | 2 | 379 | 7322967 | |
| FQ216550.1 | - | 2 | 378 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtpbp8| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025015.1 | NP_001020186.1 | 2 | 379 | ||
| GenBank mRNA | BC091236.1 | - | 2 | 379 | 7322967 | |
| FQ216550.1 | - | 2 | 378 | |||